Learn More
Enzo Life Sciences VDM-11 (50mg). CAS: 313998-81-1
SDP
Catalog No. 502010474
Encompass_Preferred
Selective and potent inhibitor of the anandamide membrane transporter (AMT) (neuronal (C6 glioma cells): IC50=11µM; non-neuronal (rat RBL-2H3 cells): IC50=7.2µM) with negligible action at TRPV1 and weak action at CB1 receptor (CB1: Ki=5µM; CB2: Ki>5µM) and FAAH (IC50=3.7µM/ IC50>50µM (N18TG2 cell membranes). Alternative name: N-Arachidonoyl-4-hydroxy-2-methylaniline, N-(4-Hydroxy-2-methylphenyl)arachidonoylamide. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (1.5mg/ml) or ethanol (1mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-0474 Supplier Enzo Life Sciences Supplier No. BMLCR1110050
Selective and potent inhibitor of the anandamide membrane transporter (AMT) (neuronal (C6 glioma cells): IC50=11µM; non-neuronal (rat RBL-2H3 cells): IC50=7.2µM) with negligible action at TRPV1 and weak action at CB1 receptor (CB1: Ki=5µM; CB2: Ki>5µM) and FAAH (IC50=3.7µM/ IC50>50µM (N18TG2 cell membranes). Alternative name: N-Arachidonoyl-4-hydroxy-2-methylaniline, N-(4-Hydroxy-2-methylphenyl)arachidonoylamide. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (1.5mg/ml) or ethanol (1mg/ml). Long Term Storage: -20°C.