Learn More
BD VDRL Flocculation Antigens
Catalog No. B40764 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
5 mL
Catalog No. Quantity
B40764 5 mL
1 options
This item is not returnable. View return policy
Catalog No. B40764 Supplier BD Supplier No. 240764
This item is not returnable. View return policy
Syphilis serology reagents
  • Nontreponemal test for syphilis
  • For qualitative and semiquantitative determinations
  • Suitable for testing cerebal spinal fluid (CSF) specimens
TRUSTED_SUSTAINABILITY
Specifications
Type VDRL Flocculation Antigen
Quantity 5 mL