Learn More
BD VDRL Flocculation Antigens
Catalog No. B40765 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B40765 0.5 mL
B40764 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. B40765 Supplier BD Supplier No. 240765
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
Syphilis serology reagents
  • Nontreponemal test for syphilis
  • For qualitative and semiquantitative determinations
  • Suitable for testing cerebal spinal fluid (CSF) specimens
TRUSTED_SUSTAINABILITY
Specifications
Type VDRL Flocculation Antigen
Quantity 0.5 mL