Learn More
Apexbio Technology LLC VE-821 1232410-49-9 100mg
SDP
Supplier:  Apexbio Technology LLC A2521100
Encompass_Preferred
VE-821 (CAS 1232410-49-9) is a selective ATP-competitive inhibitor of ATR (ataxia telangiectasia and Rad3-related) kinase a critical regulator involved in DNA damage response signaling VE-821 exhibits potent ATR inhibition with a Ki value of 13 nM and minimal off-target activity toward other PI3K-related kinases such as ATM DNA-PK mTOR and PI3K In cellular models VE-821 effectively reduces phosphorylation of ATR substrates including Chk1 at Ser345 thereby sensitizing HL-60 leukemia and pancreatic cancer cell lines (e g PSN-1 MiaPaCa-2) to chemotherapy (gemcitabine) and radiation-induced cytotoxicity VE-821 is utilized as a research tool in the study of DNA damage responses and cancer vulnerabilities
Catalog No. 50-000-62977