Learn More
Apexbio Technology LLC VE-821, 10MM (IN 1ML DMSO) Cas No: 1232410-49-9 MFCD: MFCD19443686
SDP
Supplier:  Apexbio Technology LLC A25215.1
Encompass_Preferred
VE-821 is a potent, highly-selective, and ATP-competitive DNA damage response (DDR) kinase ATR inhibitor with Ki value of 13nM. There are other sizes available. Please inqury us for quote.
Catalog No. NC1530198