Learn More
Selleck Chemical LLC VE-821
SDP
Supplier:  Selleck Chemical LLC S80071G
Encompass_Preferred
VE-821(ATR inhibitor IV) is a potent and selective ATP competitive inhibitor of ATR with Ki/IC50 of 13 nM/26 nM in cell-free assays shows inhibition of H2AX phosphorylation minimal activity against PIKKs ATM DNA-PK mTOR and PI3K
Catalog No. 50-000-00440