Learn More
Medchemexpress LLC VE-821 | 1232410-49-9 | 99.7% | 368.41 | 25 MG
SDP
Catalog No. 5000363232
Encompass_Preferred
VE-821 is a potent ATP-competitive inhibitor of ATR with a Ki/IC50 of 13 nM/26 nM. It demonstrates excellent selectivity for ATR with minimal cross-reactivity against related PIKKs such as ATM, DNA-PK, mTOR, and PI3Kγ. This inhibitor also significantly enhances the sensitivity of PSN-1, MiaPaCa-2, and primary PancM pancreatic cancer cells to radiation and Gemcitabine, under both normoxic and hypoxic conditions. ATR inhibition by VE-821 leads to the inhibition of radiation-induced G2/M arrest in cancer cells.
  • Potent ATP-competitive inhibitor of ATR
  • Ki/IC50 of 13 nM/26 nM for ATR
  • Demonstrates excellent selectivity for ATR with minimal cross-reactivity against related PIKKs
  • Enhances sensitivity of pancreatic cancer cells to radiation and Gemcitabine
  • Inhibits radiation-induced G2/M arrest in cancer cells
  • Inhibits phosphorylation of Chk1 (Ser 345)
Catalog No. 50-003-63232 Supplier Medchemexpress LLC Supplier No. 25MGHY14731
VE-821 is a potent ATP-competitive inhibitor of ATR with a Ki/IC50 of 13 nM/26 nM. It demonstrates excellent selectivity for ATR with minimal cross-reactivity against related PIKKs such as ATM, DNA-PK, mTOR, and PI3Kγ. This inhibitor also significantly enhances the sensitivity of PSN-1, MiaPaCa-2, and primary PancM pancreatic cancer cells to radiation and Gemcitabine, under both normoxic and hypoxic conditions. ATR inhibition by VE-821 leads to the inhibition of radiation-induced G2/M arrest in cancer cells.
  • Potent ATP-competitive inhibitor of ATR
  • Ki/IC50 of 13 nM/26 nM for ATR
  • Demonstrates excellent selectivity for ATR with minimal cross-reactivity against related PIKKs
  • Enhances sensitivity of pancreatic cancer cells to radiation and Gemcitabine
  • Inhibits radiation-induced G2/M arrest in cancer cells
  • Inhibits phosphorylation of Chk1 (Ser 345)