Learn More
TARGETMOL CHEMICALS INC VE-821 25MG
SDP
Catalog No. 502577018
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. VE-821 (ATR Inhibitor IV) is a selective ATP competitive inhibitor of ATR( Ki/IC50 13/26 nM in cell-free assays). purity: 99%
Catalog No. 50-257-7018 Supplier TARGETMOL CHEMICALS INC Supplier No. T303225MG
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. VE-821 (ATR Inhibitor IV) is a selective ATP competitive inhibitor of ATR( Ki/IC50 13/26 nM in cell-free assays). purity: 99%