Learn More
Apexbio Technology LLC VE-822, 1232416-25-9, MFCD27976794, 10 mg
SDP
Supplier:  Apexbio Technology LLC B138310
Encompass_Preferred
MF: C24H25N5O3S, MW: 463.55, Solubility: 50mg/mL in DMSO. VE-822 is an ATR inhibitor with an IC50 value of 0.019 uM. It is a close analog of VE-821 with a marked increase in potency against ATR.
Catalog No. 50-101-5254