Learn More
Medchemexpress LLC VE-Cadherin Mouse 10ug
SDP
Catalog No. 5000180256
Encompass_Preferred
VE-cadherin is a calcium-dependent glycoprotein critical for mediating cell adhesion and coordinating morphogenetic events during development Lack of VE-cadherin in mice leads to uterine death due to increased endothelial cell apoptosis leading to vascular insufficiency VE-Cadherin Protein Mouse (HEK293 His) is the recombinant mouse-derived VE-Cadherin protein expressed by HEK293 with C-10 His labeled tag
Catalog No. 50-001-80256 Supplier Medchemexpress LLC Supplier No. HYP7355910UG
VE-cadherin is a calcium-dependent glycoprotein critical for mediating cell adhesion and coordinating morphogenetic events during development Lack of VE-cadherin in mice leads to uterine death due to increased endothelial cell apoptosis leading to vascular insufficiency VE-Cadherin Protein Mouse (HEK293 His) is the recombinant mouse-derived VE-Cadherin protein expressed by HEK293 with C-10 His labeled tag