Learn More
Miele, Inc. VE P 2000 Cartridge Charged 1
Catalog No. 502870761
Encompass_Preferred
VE P 2000 Cartridge, Charged 12.5L. Resin capacity: 4,000L at 150µs/cm H410 Ø230mm
Catalog No. 50-287-0761 Supplier Miele, Inc. Supplier No. 9773410
VE P 2000 Cartridge, Charged 12.5L. Resin capacity: 4,000L at 150µs/cm H410 Ø230mm