Learn More
Miele, Inc. VE P 2800 Cartridge Charged 1
Catalog No. 502870745
Encompass_Preferred
VE P 2800 Cartridge, Charged 19L. Resin capacity: 5,600L at 150µs/cm H570 Ø230mm
Catalog No. 50-287-0745 Supplier Miele, Inc. Supplier No. 9782560
VE P 2800 Cartridge, Charged 19L. Resin capacity: 5,600L at 150µs/cm H570 Ø230mm