Learn More
Selleck Chemical LLC VE-821 10mg 1232410-49-9
SDP
Supplier:  Selleck Chemical LLC S800710MG
Encompass_Preferred
VE-821 is a potent and selective ATP competitive inhibitor of ATR with Ki/IC50 of 13 nM/26 nM in cell-free assays, shows inhibition of H2AX phosphorylation, minimal activity against PIKKs ATM, DNA-PK, mTOR and PI3K?. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0604741