Learn More
Medchemexpress LLC Vebicorvir | 2090064-66-5 | 99.7% | 467.44 | 50 MG
SDP
Catalog No. 5000343281
Encompass_Preferred
Vebicorvir is a first-generation inhibitor of the hepatitis B virus (HBV) core protein. It effectively suppresses the formation of covalently closed circular DNA (cccDNA) in de novo infection models, with EC50 values ranging from 1.84 μM to 7.3 μM. This compound inhibits pgRNA, HBeAg, and HBsAg production.
  • First-generation hepatitis B virus (HBV) core protein inhibitor.
  • Suppresses cccDNA formation in de novo infection models.
  • Inhibits pgRNA, HBeAg, and HBsAg production.
Catalog No. 50-003-43281 Supplier Medchemexpress LLC Supplier No. HY10919550MG
Vebicorvir is a first-generation inhibitor of the hepatitis B virus (HBV) core protein. It effectively suppresses the formation of covalently closed circular DNA (cccDNA) in de novo infection models, with EC50 values ranging from 1.84 μM to 7.3 μM. This compound inhibits pgRNA, HBeAg, and HBsAg production.
  • First-generation hepatitis B virus (HBV) core protein inhibitor.
  • Suppresses cccDNA formation in de novo infection models.
  • Inhibits pgRNA, HBeAg, and HBsAg production.