Learn More
Medchemexpress LLC Vebicorvir (ABI-H0731) | 2090064-66-5 | 99.7% | 467.44 | 100 MG
SDP
Catalog No. 5000343894
Encompass_Preferred
Vebicorvir (ABI-H0731) is a first-generation hepatitis B virus (HBV) core protein inhibitor. It suppresses covalently closed circular DNA (cccDNA) formation in two de novo infection models with EC50s from 1.84 μM to 7.3 μM.
  • Inhibits pgRNA, HBeAg, and HBsAg production, with EC50s of 2.68 μM, 4.95 μM, and 7.30 μM, respectively.
  • Acts as an inhibitor of pgRNA encapsidation and rcDNA synthesis.
Catalog No. 50-003-43894 Supplier Medchemexpress LLC Supplier No. HY109195100MG
Vebicorvir (ABI-H0731) is a first-generation hepatitis B virus (HBV) core protein inhibitor. It suppresses covalently closed circular DNA (cccDNA) formation in two de novo infection models with EC50s from 1.84 μM to 7.3 μM.
  • Inhibits pgRNA, HBeAg, and HBsAg production, with EC50s of 2.68 μM, 4.95 μM, and 7.30 μM, respectively.
  • Acts as an inhibitor of pgRNA encapsidation and rcDNA synthesis.