Learn More
Medchemexpress LLC Vebicorvir (ABI-H0731) | 2090064-66-5 | 99.7% | 467.44 g·mol^-1 | C19H12F3N3O4S2 | 5 MG
SDP
Catalog No. 5000264701
Encompass_Preferred
Vebicorvir (ABI-H0731) is a research-grade small-molecule inhibitor of the hepatitis B virus (HBV) core protein. It functions as a core protein allosteric modulator that suppresses formation of covalently closed circular DNA (cccDNA) in cellular infection models. For research use only.
  • Inhibits HBV core protein via allosteric modulation.
  • Suppresses cccDNA formation in cellular models (reported EC50 1.84-7.3 μM).
  • High chemical purity suitable for biochemical and cellular assays.
  • Molecular formula C19H12F3N3O4S2; molecular weight 467.44 g·mol^-1.
  • Supplied as a solid, soluble in common organic solvents.
  • For research use only; not for human or clinical use.
Catalog No. 50-002-64701 Supplier Medchemexpress LLC Supplier No. HY1091955MG
Vebicorvir (ABI-H0731) is a research-grade small-molecule inhibitor of the hepatitis B virus (HBV) core protein. It functions as a core protein allosteric modulator that suppresses formation of covalently closed circular DNA (cccDNA) in cellular infection models. For research use only.
  • Inhibits HBV core protein via allosteric modulation.
  • Suppresses cccDNA formation in cellular models (reported EC50 1.84-7.3 μM).
  • High chemical purity suitable for biochemical and cellular assays.
  • Molecular formula C19H12F3N3O4S2; molecular weight 467.44 g·mol^-1.
  • Supplied as a solid, soluble in common organic solvents.
  • For research use only; not for human or clinical use.