Learn More
Vector Laboratories VECTABOND Reagent For Tissue Section Adhesion 7 ml
SDP
Catalog No. NC9280699
Encompass
VECTABOND Reagent (SP-1800) is designed to significantly increase the adherence of both frozen and paraffin-embedded tissue sections and cell preparations to glass slides and coverslips. VECTABOND chemically modifies the glass to form a highly adherent surface, and tissue sections will remain attached even when subjected to the most extreme conditions, such as high temperature antigen unmasking techniques and in situ hybridization. VECTABOND Reagent-treated slides can be stored for long periods. VECTABOND Reagent is provided as 7 ml of concentrate that dilutes to 350 ml of working solution.
Catalog No. NC9280699 Supplier Vector Laboratories Supplier No. SP18007
VECTABOND Reagent (SP-1800) is designed to significantly increase the adherence of both frozen and paraffin-embedded tissue sections and cell preparations to glass slides and coverslips. VECTABOND chemically modifies the glass to form a highly adherent surface, and tissue sections will remain attached even when subjected to the most extreme conditions, such as high temperature antigen unmasking techniques and in situ hybridization. VECTABOND Reagent-treated slides can be stored for long periods. VECTABOND Reagent is provided as 7 ml of concentrate that dilutes to 350 ml of working solution.