Learn More
Vector Laboratories VectaFluor Excel Amplified DyLight 594, Anti-Mouse IgG Kit
SDP
Supplier:  Vector Laboratories DK2594
Encompass
The VectaFluor™ Excel Amplified DyLight 594, Anti-Mouse IgG Kit offers a non-biotin amplification method for fluorescence applications. This system uses a ready-to-use (R.T.U.) Amplifier Antibody, followed by a ready-to-use (R.T.U.) VectaFluor DyLight dye-labeled detection system (excitation 592 nm, emission 617 nm, color red). All the reagents in the kit are affinity-purified and extensively cross-adsorbed to ensure high sensitivity and low background. They are conjugated to the DyLight dyes using technology that ensures maximum labeling without compromising antibody affinity or specificity. DyLight dyes offer bright fluorescence, photostability, and pH independence. All reagents are supplied in stabilized, R.T.U. solutions for maximum convenience. Each kit contains 2.5% Normal Horse Serum (ready-to-use) for blocking, Amplifier Antibody (Goat anti-mouse IgG; ready-to-use), VectaFluor DyLight-labeled Horse Anti-Goat IgG (ready-to-use).
Catalog No. NC1115768