Learn More
Vector Laboratories Goat Anti-Rabbit, Unconjugated DK-2488
SDP
Supplier:  Vector Laboratories DK2488
Encompass
Goat Anti-Rabbit IgG Antibody (H+L), Unconjugated, 1.5 mg, is supplied in solution. This ultrapure, high-affinity antibody has been thoroughly adsorbed against serum and immunoglobulins from potentially interfering species and is ready for iodination, fluorochrome labeling, or enzyme conjugations. It can also be employed as a capture antibody in enzyme immunoassays or in other assays requiring carrier-free immunoglobulins.
Catalog No. 50-204-5643