Learn More
Vector Laboratories VectaFluor™ Excel Amplified Anti-Rabbit IgG, DyLight 594 Antibody Kit
SDP
Catalog No. 502045642
Encompass
VectaFluor Excel Amplified Fluorescent Staining System offers a non-biotin amplification method for fluorescence applications. This system uses a ready-to-use (R.T.U.) Amplifier Antibody, followed by a ready-to-use (R.T.U.) VectaFluor DyLight dye-labeled detection system. Features:High sensitivityLow backgroundVectaFluor DyLight 594 labeled anti-goat IgG antibody for detectionExcitation: 592 nmEmission: 617 nmColor: Red
Catalog No. 50-204-5642 Supplier Vector Laboratories Supplier No. DK1594
VectaFluor Excel Amplified Fluorescent Staining System offers a non-biotin amplification method for fluorescence applications. This system uses a ready-to-use (R.T.U.) Amplifier Antibody, followed by a ready-to-use (R.T.U.) VectaFluor DyLight dye-labeled detection system. Features:High sensitivityLow backgroundVectaFluor DyLight 594 labeled anti-goat IgG antibody for detectionExcitation: 592 nmEmission: 617 nmColor: Red