Learn More
Enterprise Technology Solutions Vectair Systems, Inc. Urinal Screen, Linen Breeze, 6-3/5"x6-3/5"x1/4", 10/CT, Blue
SDP
Catalog No. 5000829218
Encompass_Preferred
The Wee-Screen features interlacing strips of EVA material that form a strong lattice to make it less rigid than other screens. Linen Breeze scent produces a hint of clean linen to create a relaxing and uplifting atmosphere. Long-lasting fragrance reduces bad smells for up to 30 days. Deep bubble design prevents splashback and drain blockages. Slightly raised grip pad on the edge of the screen allows quick, easy servicing. Urinal screen fits most urinal shapes and sizes.
Catalog No. 50-008-29218 Supplier Enterprise Technology Solutions Supplier No. VTSWSCRNLIN
The Wee-Screen features interlacing strips of EVA material that form a strong lattice to make it less rigid than other screens. Linen Breeze scent produces a hint of clean linen to create a relaxing and uplifting atmosphere. Long-lasting fragrance reduces bad smells for up to 30 days. Deep bubble design prevents splashback and drain blockages. Slightly raised grip pad on the edge of the screen allows quick, easy servicing. Urinal screen fits most urinal shapes and sizes.