Learn More
Vector Laboratories VectaMount Express Mounting Medium, 60 ml
SDP
Catalog No. NC2004138
Encompass
VectaMount Express is a non-aqueous clearing and mounting medium enabling the rapid mounting of cell and tissue specimens following immunohistochemistry (IHC) staining. This novel formulation is engineered to enable mounting directly following staining, saving time by eliminating the need for extensive ethanol and clearing washes prior to coverslipping. Just stain your slides as per your usual workflow, briefly wash in isopropyl alcohol, and coverslip. Features: Non-aqueous clearing and mounting medium for IHC-stained slides. Eliminates the need for extensive ethanol washes and solvent-based clearing agents (e.g., xylene).Rapid drying formula for fast visualization of stained samples. Preserves staining for at least 18 months at room temperature.
Catalog No. NC2004138 Supplier Vector Laboratories Supplier No. H570060
VectaMount Express is a non-aqueous clearing and mounting medium enabling the rapid mounting of cell and tissue specimens following immunohistochemistry (IHC) staining. This novel formulation is engineered to enable mounting directly following staining, saving time by eliminating the need for extensive ethanol and clearing washes prior to coverslipping. Just stain your slides as per your usual workflow, briefly wash in isopropyl alcohol, and coverslip. Features: Non-aqueous clearing and mounting medium for IHC-stained slides. Eliminates the need for extensive ethanol washes and solvent-based clearing agents (e.g., xylene).Rapid drying formula for fast visualization of stained samples. Preserves staining for at least 18 months at room temperature.