Learn More
Vector Laboratories Vibrance Antifade Mounting Medium, 2ml
SDP
Catalog No. NC1601053
Encompass
VECTASHIELD Vibrance is an aqueous mountant that sets at room temperature. Slides are viewable within one hour after mounting and can be stored at room temperature for extended periods, without loss of signal intensity, retraction of media or bubble formation. VECTASHIELD Vibrance Antifade Mounting Media provides excellent protection against fading across the visible spectrum, even under far-red wavelengths. It are compatible with most commercially available fluorophores. Features: Superior antifade/anti-photobleaching properties across the spectrum, Compatibility with commonly used fluorophores, View sections one hour after mounting, No tone or autofluorescent background, even after curing
Catalog No. NC1601053 Supplier Vector Laboratories Supplier No. H17002
VECTASHIELD Vibrance is an aqueous mountant that sets at room temperature. Slides are viewable within one hour after mounting and can be stored at room temperature for extended periods, without loss of signal intensity, retraction of media or bubble formation. VECTASHIELD Vibrance Antifade Mounting Media provides excellent protection against fading across the visible spectrum, even under far-red wavelengths. It are compatible with most commercially available fluorophores. Features: Superior antifade/anti-photobleaching properties across the spectrum, Compatibility with commonly used fluorophores, View sections one hour after mounting, No tone or autofluorescent background, even after curing