Learn More
Vector Laboratories VECTASHIELD Antifade Mounting Medium with Propidium Iodide (PI)
SDP
Catalog No. NC1661736
Encompass
VECTASHIELD Antifade Mounting Medium with PI, prevents rapid photobleaching of fluorescent proteins and fluorescent dyes. PI is a reddish nuclear counterstain with a broad excitation range at around 530 nm and emission at about 615 nm. Features: Ideal refractive index (1.45)Ready-to-use - from the refrigerator to the benchtop, No warming necessary, Can be stored without sealing for long term analysis Non-hardening formulation
Catalog No. NC1661736 Supplier Vector Laboratories Supplier No. H130010
VECTASHIELD Antifade Mounting Medium with PI, prevents rapid photobleaching of fluorescent proteins and fluorescent dyes. PI is a reddish nuclear counterstain with a broad excitation range at around 530 nm and emission at about 615 nm. Features: Ideal refractive index (1.45)Ready-to-use - from the refrigerator to the benchtop, No warming necessary, Can be stored without sealing for long term analysis Non-hardening formulation