Learn More
Vector Laboratories VECTASHIELD PLUS Antifade Mounting Medium, 2ml
SDP
Catalog No. 502045660
Encompass
VECTASHIELD PLUS Antifade Mounting Medium is a unique, stable formula for preserving fluorescence. This medium inhibits rapid photobleaching of fluorescent proteins and fluorescent dyes. Features: No inherent toning or background - Superior signal retention across the spectrum from blue to far red - Mounted sections can be viewed in 1 hour - Available in two convenient sizes - Superior non-setting formulation -
Catalog No. 50-204-5660 Supplier Vector Laboratories Supplier No. H19002
VECTASHIELD PLUS Antifade Mounting Medium is a unique, stable formula for preserving fluorescence. This medium inhibits rapid photobleaching of fluorescent proteins and fluorescent dyes. Features: No inherent toning or background - Superior signal retention across the spectrum from blue to far red - Mounted sections can be viewed in 1 hour - Available in two convenient sizes - Superior non-setting formulation -