Learn More
Vector Laboratories VECTASTAIN ABC-AP Staining Kit (Alkaline Phosphatase, Rabbit IgG)
SDP
Catalog No. NC9617531
Encompass
With more than 50,000 citations, Vector Laboratories ABC kits are trusted by immunohistochemistry (IHC) and immunocytochemistry (ICC) laboratories to provide reliable results. All of the VECTASTAIN kits are based on the versatile avidin-biotin complex (ABC) interaction, and different kit variations can be used to accommodate a wide array of primary antibody and tissue species. This kit uses alkaline phosphatase (AP) as a detection enzyme and includes a secondary antibody that targets rabbit IgG primary antibodies. The ABC complex, once formed, is stable and can be used for several days after preparation. VECTASTAIN ABC-AP KIT (Rabbit IgG) consists of 3 ml normal goat serum, 1 ml biotinylated goat anti-rabbit IgG secondary antibody, 2 ml Reagent A (avidin) and 2 ml Reagent B (biotinylated AP). Stains 1000-2000 sections.
Catalog No. NC9617531 Supplier Vector Laboratories Supplier No. AK5001
With more than 50,000 citations, Vector Laboratories ABC kits are trusted by immunohistochemistry (IHC) and immunocytochemistry (ICC) laboratories to provide reliable results. All of the VECTASTAIN kits are based on the versatile avidin-biotin complex (ABC) interaction, and different kit variations can be used to accommodate a wide array of primary antibody and tissue species. This kit uses alkaline phosphatase (AP) as a detection enzyme and includes a secondary antibody that targets rabbit IgG primary antibodies. The ABC complex, once formed, is stable and can be used for several days after preparation. VECTASTAIN ABC-AP KIT (Rabbit IgG) consists of 3 ml normal goat serum, 1 ml biotinylated goat anti-rabbit IgG secondary antibody, 2 ml Reagent A (avidin) and 2 ml Reagent B (biotinylated AP). Stains 1000-2000 sections.