Learn More
Vector Laboratories VECTASTAIN ABC-AP, Rabbit IgG AK-5200
SDP
Supplier:  Vector Laboratories AK5200
Encompass
VECTASTAIN ABC-AP Kit, Alkaline Phosphatase (Rabbit IgG), 1 Kit. Based on the versatile avidin-biotin complex interaction, the VECTASTAIN ABC AP Staining system is modular and, with Vector Laboratories' selection of secondary antibodies, can accommodate a wide array of primary antibody and tissue species. ABC kits are economical and are a staple product in IHC and ICC laboratories. The VECTASTAIN ABC Reagent can be used to detect any biotinylated molecule. This is the original avidin/biotin ABC complex formation with a biotinylated anti-rabbit IgG secondary antibody. Features: Alkaline phosphatase enzyme detection system, Avidin/biotin technology, High sensitivity/low background, Preferred for tissues that have high endogenous peroxidase activity, Additional substrate choices, Used as a second label in multiple antigen labeling applications. Kit Contents: 3 ml Normal Goat Serum, 1 ml Biotinylated Goat Anti-Rabbit IgG, 2 ml Reagent A (Avidin DH) plus 2 ml Reagent B (Biotinylated AP).
Catalog No. NC9764322