Learn More
R&D Systems™ VECTASTAIN™ ELITE™ ABC anti-Mouse IgG HRP Immunodetection Kit

Specifications
Specifications
| Target Molecule | VECTASTAIN ELITE ABC anti-Mouse IgG |
| Quantity | 1 Kit |
| Content And Storage | Store at 4°C. Do not freeze. |
| For Use With (Application) | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, ISH, ELISpot |
| Product Line | Novus |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.