Learn More
Vector Laboratories VECTASTAIN Elite ABC HRP Kit (Peroxidase, Goat IgG) Kit
SDP
Catalog No. NC9461324
Encompass
The highly stable bond between avidin (A) and biotin (B) can be used to form a complex (ABC) between a biotinylated protein or other target and an avidin-conjugated detection enzyme. With more than 50,000 citations, Vector Laboratories ABC kits are trusted by immunohistochemistry (IHC) and immunocytochemistry (ICC) laboratories. Different variations of the ABC kit system can accommodate a wide array of targets and tissue species. This VECTASTAIN® Elite® ABC-HRP Kit (Peroxidase, Anti-Goat IgG) features a small, highly active avidin/biotin-HRP complex for superior target access and sensitivity. Typical sensitivity is 5X higher than the original ABC kits. The kit includes 1 ml of biotinylated, affinity-purified rabbit anti-goat IgG antibody, 2 ml Reagent A (avidin) and 2 ml Reagent B (biotinylated HRP) and 3 ml normal rabbit serum. Stains 500-1000 sections.
Catalog No. NC9461324 Supplier Vector Laboratories Supplier No. PK6105
The highly stable bond between avidin (A) and biotin (B) can be used to form a complex (ABC) between a biotinylated protein or other target and an avidin-conjugated detection enzyme. With more than 50,000 citations, Vector Laboratories ABC kits are trusted by immunohistochemistry (IHC) and immunocytochemistry (ICC) laboratories. Different variations of the ABC kit system can accommodate a wide array of targets and tissue species. This VECTASTAIN® Elite® ABC-HRP Kit (Peroxidase, Anti-Goat IgG) features a small, highly active avidin/biotin-HRP complex for superior target access and sensitivity. Typical sensitivity is 5X higher than the original ABC kits. The kit includes 1 ml of biotinylated, affinity-purified rabbit anti-goat IgG antibody, 2 ml Reagent A (avidin) and 2 ml Reagent B (biotinylated HRP) and 3 ml normal rabbit serum. Stains 500-1000 sections.