Learn More
Vector Laboratories VECTASTAIN Universal Quick HRP Kit, Peroxidase (Concentrate)
SDP
Catalog No. 502045695
Encompass
he VECTASTAIN Universal Quick Kit is designed for rapid immunohistochemical staining of tissue sections. The kit relies on a proprietary preformed peroxidase-streptavidin complex to achieve outstanding sensitivity with short incubation times.
Features:
• Rapid protocol: Staining in less than 20 minutes following primary antibody incubation. Working solutions can be used immediately after dilution.
• High sensitivity, low background.
• Biotinylated, Universal Pan-Specific secondary antibody recognizes mouse, rabbit, and goat primary antibodies as well as those from related species such as rat, bovine, and sheep.
• Concentrated format: Reagents are supplied as liquid concentrates enabling flexibility in dilution and application while maximizing cost-savings per assay.
Catalog No. 50-204-5695 Supplier Vector Laboratories Supplier No. PK8800
he VECTASTAIN Universal Quick Kit is designed for rapid immunohistochemical staining of tissue sections. The kit relies on a proprietary preformed peroxidase-streptavidin complex to achieve outstanding sensitivity with short incubation times.
Features:
• Rapid protocol: Staining in less than 20 minutes following primary antibody incubation. Working solutions can be used immediately after dilution.
• High sensitivity, low background.
• Biotinylated, Universal Pan-Specific secondary antibody recognizes mouse, rabbit, and goat primary antibodies as well as those from related species such as rat, bovine, and sheep.
• Concentrated format: Reagents are supplied as liquid concentrates enabling flexibility in dilution and application while maximizing cost-savings per assay.