Learn More
Vector Laboratories VECTOR Black Alkaline Phosphatase (AP) Substrate Kit 200 ml
SDP
Catalog No. NC9703722
Encompass
VECTOR Black Alkaline Phosphatase AP Substrate Kit forms a brown-black precipitate that can be dehydrated, cleared, and permanently mounted. This chromogen can be used in immunohistochemical or blotting applications. The stock solutions in this kit are provided in convenient dropper bottles, in sufficient volume for 200 ml working solution.
Catalog No. NC9703722 Supplier Vector Laboratories Supplier No. SK5200
VECTOR Black Alkaline Phosphatase AP Substrate Kit forms a brown-black precipitate that can be dehydrated, cleared, and permanently mounted. This chromogen can be used in immunohistochemical or blotting applications. The stock solutions in this kit are provided in convenient dropper bottles, in sufficient volume for 200 ml working solution.