Learn More
Cell Biolabs Inc pAAV-MCS Expression Vector, 10 µg
Supplier:  Cell Biolabs Inc VPK410
Encompass_Preferred
Cell Biolabs' pAAV-MCS Expression Vector contains a CMV promoter ahead of the multiple cloning sites. Clone your gene into the pAAV-MCS Expression Vector and then co-transfect with AAV packaging plasmids into 293 cells to generate recombinant AAV. The cloning capacity is 3.0 kb.
Catalog No. 50-673-813