Learn More
Vector Laboratories VECTOR Hematoxylin QS 100 ml
SDP
Catalog No. NC9788954
Encompass
Vector Hematoxylin QS is a modification of Mayer's Hematoxylin and was developed especially for immunocytochemistry. It provides crisp blue nuclear staining. This formula is ready-to-use without filtration, requires no separate bluing step, and stains in less than 45 seconds. Vector Hematoxylin QS contains no mercury and provides excellent color contrast with most commonly used peroxidase and alkaline phosphatase substrates. This counterstain is suitable for use with non-aqueous and aqueous mountants.
Catalog No. NC9788954 Supplier Vector Laboratories Supplier No. H3404100
Vector Hematoxylin QS is a modification of Mayer's Hematoxylin and was developed especially for immunocytochemistry. It provides crisp blue nuclear staining. This formula is ready-to-use without filtration, requires no separate bluing step, and stains in less than 45 seconds. Vector Hematoxylin QS contains no mercury and provides excellent color contrast with most commonly used peroxidase and alkaline phosphatase substrates. This counterstain is suitable for use with non-aqueous and aqueous mountants.