Learn More
Invivogen Human kappa, pFUSE2ss-CLIg-hK, 20 ug
SDP
Catalog No. NC1833775
Encompass
pFUSE2ss-CLIg-hK additionally features the IL2 signal sequence to allow the secretion of recombinant antibodies generated using fragment antigen-binding (Fab) or single-chain variable fragment (scFv) fragments selected from phage display libraries that lack a signal sequence.
Catalog No. NC1833775 Supplier Invivogen Supplier No. PFUSE2SSHCLK
pFUSE2ss-CLIg-hK additionally features the IL2 signal sequence to allow the secretion of recombinant antibodies generated using fragment antigen-binding (Fab) or single-chain variable fragment (scFv) fragments selected from phage display libraries that lack a signal sequence.