Learn More
Cell Biolabs Inc pSMPUW Promoterless Lentiviral Expression Vector, 10 µg
Catalog No. 50673760
Encompass_Preferred
Cell Biolabs' pSMPUW Promoterless Lentiviral Expression Vector allows you to clone a gene along with its optimal promoter up to 9.4 kb (total insert capacity). This vector can be used in conjunction with any second or third generation lentiviral packaging system, but it produces the highest viral titers when used with the ViraSafe™ Lentivirus Packaging System (Ecotropic or Pantropic).
Catalog No. 50-673-760 Supplier Cell Biolabs Inc Supplier No. VPK211
Cell Biolabs' pSMPUW Promoterless Lentiviral Expression Vector allows you to clone a gene along with its optimal promoter up to 9.4 kb (total insert capacity). This vector can be used in conjunction with any second or third generation lentiviral packaging system, but it produces the highest viral titers when used with the ViraSafe™ Lentivirus Packaging System (Ecotropic or Pantropic).