Learn More
Vector Laboratories Vector M.O.M. (Mouse-On-Mouse) Immunodetection Kit
SDP
Catalog No. NC9685284
Encompass
Vector M.O.M. Kits contain a proprietary Mouse IgG Blocking Reagent that greatly reduces endogenous mouse IgG background when staining mouse primary antibodies on mouse tissue. The Vector M.O.M. Kit eliminates this problem, with clear, crisp specific staining of the antigens of interest. The kit is based on the simple and easy to follow VECTASTAIN ABC reagent protocols; no tedious calculations or antibody pre-binding steps are required. Each kit contains a proprietary Mouse IgG Blocking Reagent, M.O.M. Biotinylated Anti-Mouse Ig Reagent, M.O.M. Protein Concentrate for general protein blocking, and VECTASTAIN Elite ABC Reagent. Vector M.O.M. Immunodetection Kits are useful in any mouse-on-mouse application, such as studies in genetically engineered mice, including transgenic and knock-out models, and mouse xenografts.
Catalog No. NC9685284 Supplier Vector Laboratories Supplier No. PK2200
Vector M.O.M. Kits contain a proprietary Mouse IgG Blocking Reagent that greatly reduces endogenous mouse IgG background when staining mouse primary antibodies on mouse tissue. The Vector M.O.M. Kit eliminates this problem, with clear, crisp specific staining of the antigens of interest. The kit is based on the simple and easy to follow VECTASTAIN ABC reagent protocols; no tedious calculations or antibody pre-binding steps are required. Each kit contains a proprietary Mouse IgG Blocking Reagent, M.O.M. Biotinylated Anti-Mouse Ig Reagent, M.O.M. Protein Concentrate for general protein blocking, and VECTASTAIN Elite ABC Reagent. Vector M.O.M. Immunodetection Kits are useful in any mouse-on-mouse application, such as studies in genetically engineered mice, including transgenic and knock-out models, and mouse xenografts.