Learn More
Vector Laboratories VECTASHIELD Antifade Mounting Medium With DAPI 10 ml
SDP
Catalog No. NC9524612
Encompass
VECTASHIELD Antifade Mounting Medium prevents rapid photobleaching of fluorescent proteins and dyes. It remains a semi-liquid on the slide. If desired, coverslips can be sealed around the perimeter with nail polish or a plastic sealant. Mounted slides should be stored in the dark at 4 degrees C. VECTASHIELD Mounting Medium is compatible with a wide array of fluorochromes, enzymatic substrates, and fluorescent proteins. The refractive index is 1.45. DAPI is a counterstain that binds to nuclei and causes them to fluoresce with a blue color. This is useful for Fluorescence In Situ Hybridization (FISH) assays and other techniques where a general orientation to the location of nuclei and cells is helpful.
Catalog No. NC9524612 Supplier Vector Laboratories Supplier No. H120010
VECTASHIELD Antifade Mounting Medium prevents rapid photobleaching of fluorescent proteins and dyes. It remains a semi-liquid on the slide. If desired, coverslips can be sealed around the perimeter with nail polish or a plastic sealant. Mounted slides should be stored in the dark at 4 degrees C. VECTASHIELD Mounting Medium is compatible with a wide array of fluorochromes, enzymatic substrates, and fluorescent proteins. The refractive index is 1.45. DAPI is a counterstain that binds to nuclei and causes them to fluoresce with a blue color. This is useful for Fluorescence In Situ Hybridization (FISH) assays and other techniques where a general orientation to the location of nuclei and cells is helpful.