Learn More
Vector Laboratories VectaFluor Duet Kit (DyLight 488 X-Rabbit IgG/DyLight 594 X-Mouse IgG)
SDP
Catalog No. NC1427091
Encompass
This VectaFluor Duet Immunofluorescence Double Labeling Kit detects mouse and rabbit primary antibodies in a single step using horse secondary antibodies labeled with green and red DyLight fluorescent dyes. The affinity-purified, extensively cross-adsorbed secondary antibodies are conjugated to DyLight dyes in a manner that ensures maximum labeling without compromising antibody affinity or specificity. VectaFluor Duet is compatible with fluorescence staining of cells and tissues. Each kit contains 2.5% Normal Horse Serum (ready-to-use) and 15 ml VectaFluor Duet Reagent [DyLight 488 Anti-Rabbit IgG and DyLight 594 Anti-Mouse IgG cocktail (rabbit-green, mouse-red)] made in horse and ready-to-use in 5% NHS.
Catalog No. NC1427091 Supplier Vector Laboratories Supplier No. DK8818
This VectaFluor Duet Immunofluorescence Double Labeling Kit detects mouse and rabbit primary antibodies in a single step using horse secondary antibodies labeled with green and red DyLight fluorescent dyes. The affinity-purified, extensively cross-adsorbed secondary antibodies are conjugated to DyLight dyes in a manner that ensures maximum labeling without compromising antibody affinity or specificity. VectaFluor Duet is compatible with fluorescence staining of cells and tissues. Each kit contains 2.5% Normal Horse Serum (ready-to-use) and 15 ml VectaFluor Duet Reagent [DyLight 488 Anti-Rabbit IgG and DyLight 594 Anti-Mouse IgG cocktail (rabbit-green, mouse-red)] made in horse and ready-to-use in 5% NHS.