Learn More
Vector Laboratories VECTASHIELD Antifade Mounting Medium 10 ml
SDP
Catalog No. NC9265087
Encompass
VECTASHIELD Antifade Mounting Medium prevents rapid photobleaching of fluorescent proteins and dyes. It does not solidify, but remains a semi-liquid on the slide and can be stored without sealing. If desired, coverslips can be sealed around the perimeter with nail polish or a plastic sealant. Mounted slides should be stored at 4 degrees C, protected from light. VECTASHIELD Mounting Medium is compatible with a wide array of fluorochromes, enzymatic substrates, and fluorescent proteins, including many of the fluorophores used in super resolution methods like STORM, STED, and 3D-SIMM imaging, extending their lifetime and enhancing their photo-switching characteristics. (Olivier, N. et al., 2013, Biomedical Optics Express, Vol. 4 No. 6, pp. 885-899). The refractive index is 1.45. Ships at Ambient Temperature and Once received the storage temperature is 2-8 °C.
Catalog No. NC9265087 Supplier Vector Laboratories Supplier No. H100010
VECTASHIELD Antifade Mounting Medium prevents rapid photobleaching of fluorescent proteins and dyes. It does not solidify, but remains a semi-liquid on the slide and can be stored without sealing. If desired, coverslips can be sealed around the perimeter with nail polish or a plastic sealant. Mounted slides should be stored at 4 degrees C, protected from light. VECTASHIELD Mounting Medium is compatible with a wide array of fluorochromes, enzymatic substrates, and fluorescent proteins, including many of the fluorophores used in super resolution methods like STORM, STED, and 3D-SIMM imaging, extending their lifetime and enhancing their photo-switching characteristics. (Olivier, N. et al., 2013, Biomedical Optics Express, Vol. 4 No. 6, pp. 885-899). The refractive index is 1.45. Ships at Ambient Temperature and Once received the storage temperature is 2-8 °C.