Learn More
Apexbio Technology LLC Vecuronium Bromide 50700-72-6 25mg
SDP
Catalog No. 5000061884
Encompass_Preferred
Vecuronium Bromide (CAS 50700-72-6) is a steroidal neuromuscular-blocking agent that acts by competitively antagonizing acetylcholine at nicotinic receptors located on the neuromuscular junction By inhibiting acetylcholine-mediated signal transmission this compound induces skeletal muscle relaxation and reversible paralysis without depolarizing muscle cells In biomedical research Vecuronium Bromide serves as an important pharmacological tool for studying neuromuscular transmission acetylcholine receptor pharmacodynamics and muscle contraction mechanisms
Catalog No. 50-000-61884 Supplier Apexbio Technology LLC Supplier No. A522025
Vecuronium Bromide (CAS 50700-72-6) is a steroidal neuromuscular-blocking agent that acts by competitively antagonizing acetylcholine at nicotinic receptors located on the neuromuscular junction By inhibiting acetylcholine-mediated signal transmission this compound induces skeletal muscle relaxation and reversible paralysis without depolarizing muscle cells In biomedical research Vecuronium Bromide serves as an important pharmacological tool for studying neuromuscular transmission acetylcholine receptor pharmacodynamics and muscle contraction mechanisms