Learn More
Medchemexpress LLC Vecuronium bromide | 50700-72-6 | MFCD00867762 | 99.9% | 100 MG
SDP
Catalog No. 5000450862
Encompass_Preferred
Vecuronium bromide is a non-depolarizing neuromuscular blocking agent. It also functions as a nicotinic acetylcholine receptor (nAChR) inhibitor and a muscle relaxant. This compound can be used for pre-surgical anesthesia and is intended for research use only.
  • Non-depolarizing neuromuscular blocking agent
  • Inhibits nicotinic acetylcholine receptors (nAChR)
  • Acts as a muscle relaxant
  • Can be used for pre-surgical anesthesia
  • Suitable for research applications only
Catalog No. 50-004-50862 Supplier Medchemexpress LLC Supplier No. HYB0118A100MG
Vecuronium bromide is a non-depolarizing neuromuscular blocking agent. It also functions as a nicotinic acetylcholine receptor (nAChR) inhibitor and a muscle relaxant. This compound can be used for pre-surgical anesthesia and is intended for research use only.
  • Non-depolarizing neuromuscular blocking agent
  • Inhibits nicotinic acetylcholine receptors (nAChR)
  • Acts as a muscle relaxant
  • Can be used for pre-surgical anesthesia
  • Suitable for research applications only