Learn More
Medchemexpress LLC HY-B0118A 50mg Medchemexpress, Vecuronium (bromide) CAS:50700-72-6 Purity:>98%
SDP
Catalog No. 502082368
Encompass_Preferred
Medchemexpress, HY-B0118A 50mg Vecuronium (bromide) CAS:50700-72-6 Vecuronium bromide (ORG NC 45) is a nondepolarizing neuromuscular blocking agent of intermediate duration. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2368 Supplier Medchemexpress LLC Supplier No. HYB0118A50MG
Medchemexpress, HY-B0118A 50mg Vecuronium (bromide) CAS:50700-72-6 Vecuronium bromide (ORG NC 45) is a nondepolarizing neuromuscular blocking agent of intermediate duration. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.