Learn More
Cayman Chemical Vecuronium (bromide), Purity: ≥98%, CAS: 50700-72-6, 50 mg
SDP
Catalog No. 502969929
Encompass_Preferred
A non-depolarizing muscle relaxant that competitively blocks cholinergic receptors at the motor end plate of the neuromuscular junction, inducing temporary paralysis
Catalog No. 50-296-9929 Supplier Cayman Chemical Supplier No. 1560350 MG
A non-depolarizing muscle relaxant that competitively blocks cholinergic receptors at the motor end plate of the neuromuscular junction, inducing temporary paralysis