Learn More
Medchemexpress LLC HY-P9911 1mg Medchemexpress, Vedolizumab CAS:943609-66-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP99111MG
Encompass_Preferred
Medchemexpress, HY-P9911 1mg Vedolizumab CAS:943609-66-3 Vedolizumab is a humanized monoclonal antibody that targets the α4β7 integrin for the treatment of ulcerative colitis and Crohn's disease. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4397