Learn More
Perkin Elmer US LLC VeeMAX Flat Plate - Ge, 45°
Catalog No. 502113246
Encompass_Preferred
The VeeMAX III with ATR accessory offers continuous variable angle of incidence and a variety of crystal plates to selectively control the depth of penetration of the IR beam into the sample. The design allows for constant purge, which is not broken upon sample loading, and an integrated polarizer slot. ATR applications include the study of layered samples, coatings, release agents, monolayers on silicon and chemical migration studies. An electrochemical cell add-on and a temperature control option are also available. Compatible with the Spectrum One/100/400/65 and the Spectrum 3/Frontier spectrometers.
Catalog No. 50-211-3246 Supplier Perkin Elmer US LLC Supplier No. L1272452
The VeeMAX III with ATR accessory offers continuous variable angle of incidence and a variety of crystal plates to selectively control the depth of penetration of the IR beam into the sample. The design allows for constant purge, which is not broken upon sample loading, and an integrated polarizer slot. ATR applications include the study of layered samples, coatings, release agents, monolayers on silicon and chemical migration studies. An electrochemical cell add-on and a temperature control option are also available. Compatible with the Spectrum One/100/400/65 and the Spectrum 3/Frontier spectrometers.