Learn More
Glass Expansion VeeSpray Ceramic Nebulizer, Includes STT-16-75 Sample Tubing 0.75mm ID x 1/16 OD (750mm length), and 70-803-1516 Type 13 DC Fitting with Ratchet Nebulizer connector.
SDP
Catalog No. 501907533
Encompass_Preferred
VeeSpray U-Series Nebulizer, includes UniFit Connector with 0.75mm ID x 1.6mm OD x 750mm long tubing (1 pc) & universal gas connection kit.Optimum nebulizer gas flow rate is 1.0L/min & sample flow rate best between 1.5 - 2.5mL/min, must be pumped. Backpressure should read ~40 psi at 1.0L/min gas flow rate. VeeSpray is a V-groove design made from ceramic, ideal for greatest resistance to large particulates, high TDS & HF. Tolerates up to 30% TDS & 300um particulates, providing a robust, virtually unblockable nebulizer. Replacement parts include P/Ns STT-16-75, 70-803-1350, 70-803-0780, 70-803-1485 & 502702994.Recommend nebulizer cleaning solution is Fluka RBS25 (P/N NC9000117). When nebulizer is not in use, store in the provided hard plastic case.Consider upgrading to our Direct Connect (DC) VeeSpray to take advantage of the cost-effective design improvements.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7533 Supplier Glass Expansion Supplier No. ARG1CV6
VeeSpray U-Series Nebulizer, includes UniFit Connector with 0.75mm ID x 1.6mm OD x 750mm long tubing (1 pc) & universal gas connection kit.Optimum nebulizer gas flow rate is 1.0L/min & sample flow rate best between 1.5 - 2.5mL/min, must be pumped. Backpressure should read ~40 psi at 1.0L/min gas flow rate. VeeSpray is a V-groove design made from ceramic, ideal for greatest resistance to large particulates, high TDS & HF. Tolerates up to 30% TDS & 300um particulates, providing a robust, virtually unblockable nebulizer. Replacement parts include P/Ns STT-16-75, 70-803-1350, 70-803-0780, 70-803-1485 & 502702994.Recommend nebulizer cleaning solution is Fluka RBS25 (P/N NC9000117). When nebulizer is not in use, store in the provided hard plastic case.Consider upgrading to our Direct Connect (DC) VeeSpray to take advantage of the cost-effective design improvements.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001