Learn More
Epredia™ Vega 6 Hopper Laser Cassette Printer with DUO and Total Care Package
Catalog No. 21403521 Shop All Epredia Products
Change view
Click to view available options
Capacity:
480 Cassettes
Catalog No. Capacity
21-403-521 480 Cassettes
1 options
Catalog No. 21-403-521 Supplier Epredia™ Supplier No. 8000041B
Add to Cart
Add to Cart
Produces high quality prints combined with a patented chemical additive on the cassette to deliver consistency and efficiency. Built in interface connectivity ensures LIS/LIM compatibility.
  • Laser printing technology for permanent and durable cassette marking
  • Supports alphanumeric characters and barcode formats, including linear, bidimensional, and ASCII codes
  • High-resolution printing at 600 dpi for clear and precise output
  • Print speed of approximately 5 seconds per cassette
  • Designed for high-throughput environments with automated cassette feeding
  • Suitable for laboratories with high-volume printing requirements
  • No requirement for consumable components such as print heads or ribbons
  • Reduced maintenance requirements and lower operating costs
  • Available in two configurations: single cassette hopper (NOVA) and six-cassette hoppers (VEGA)
  • Supports printing on multiple cassette colors and styles
  • Compact design for efficient use of laboratory space
  • Low noise operation during printing
  • Fully automated operation for streamlined workflow
  • Includes a 1-year base warranty with an additional 2-year service plan option
  • Service plan covers parts, labor, and annual preventive maintenance
  • Electrical requirements: 100/240 V, 50/60 Hz
  • Complies with IEC 61010-2-101:2015, IEC 61010-1:2010, IEC 61326-1:2012, IEC 61326-2-6:2013 standards
Specifications
Capacity 480 Cassettes
Includes DUO and Total Care Package
Type Laser Printer
Voltage 100/240 V
Color Gray