Learn More
Epredia™ Vega 8 Hopper Laser Cassette Printer with PC
Catalog No. VEGA06FTPC Shop All Epredia Products
Change view
Click to view available options
Capacity:
640 Cassettes
Catalog No. Capacity
VEGA06FTPC 640 Cassettes
1 options
Catalog No. VEGA06FTPC Supplier Epredia™ Supplier No. VEGA06FTPCDI
Add to Cart
Add to Cart
The technology produces high quality prints combined with a patented chemical additive on the cassette to deliver consistency and efficiency.
  • High Printing Quality
  • Epredia™ laser printers create sustainable and indelible printed information where patient data can be identified accurately throughout the lifetime of the cassette supporting laboratories and patients.
  • Character Types: Alphanumeric
  • Barcode Types: Linear, Bidimensional, ASCII Codes High Throughput
  • Also incorporate a highly efficient technology helping labs with higher printing demands utilize the laser printing capabilities and automated cassette feeder feature to achieve high throughput and faster Turn-around times. Low Operations Cost
  • This does not require replacement of ancillary components significantly decreasing operation costs and lowers any intensive maintenance helping lab managers from being burdened with high operation costs.
  • Features Benefits Epredia™ NOVA laser Printer is a single cassette hopper printer providing an economical option for laboratories.
  • Epredia™ VEGA™ laser printer has six cassette hoppers and supports laboratories printing a variety of cassette colors and styles.
  • Laser Technology eliminates ancillary components such as print heads ribbons and simplifies operation.
  • High resolution printing, 600 dpi, improves printing sharpness and clarity of characters allowing more characters to be embedded on cassette surface.
  • Fully automated system with fast print speed, 5 seconds per cassette, allows the user to quickly and easily print cassettes needed for a case at accessioning or grossing.
  • Laser system with cassette additive print technology ensures permanent and delible marking that are extremely durable overtime.
  • Small footprint reduces the concern of space constraint in the lab.
  • Quiet printing reduces distraction during grossing and accessioning. Total Care Package
  • 1 year base warranty and 2 additional years of total care plan
  • Plan includes parts and labor for repairs, and one preventive maintenance visit per year
  • Built in interface connectivity ensures LIS/LIM compatibility.
  • This Total Care Service Package agreement is conditional upon the Customer using only Epredia laser cassettes in the laser cassette printer. If other cassette types are used during the term of this Agreement, then this Agreement shall revert to the Premier Service coverage.
Specifications
Capacity 640 Cassettes
Type Laser Printer
Voltage 100/240 V
Color Gray