Learn More
Epredia™ Vega 8-Hopper Laser Cassette Printer
Catalog No. 23900112 Shop All Epredia Products
Change view
Click to view available options
Type:
Laser Printer
Catalog No. Type
23-900-112 Laser Printer
1 options
Catalog No. 23-900-112 Supplier Epredia™ Supplier No. 8000062
Add to Cart
Add to Cart
The Epredia™ Vega Laser Cassette Printer uses advanced technology to produce high-quality prints, featuring a patented chemical additive on the cassette for consistent and efficient performance. Built-in interface connectivity ensures compatibility with LIS/LIM systems.
Epredia™ Vega Laser Cassette Printer delivers high-quality prints with chemical-treated cassettes and LIS/LIM compatibility. It combines advanced laser technology with reliable performance to meet the demands of modern histology laboratories.
Key Features & Benefits:
  • Sustainable, indelible prints ensure accurate patient data identification throughout the cassette's lifespan.
  • Chemical additive on cassettes enhances print durability and clarity.
  • Supports alphanumeric characters and barcodes (Linear, Bidimensional, ASCII).
  • High throughput with automated cassette feeder and fast 5-second print speed.
  • 600 dpi resolution for sharp, clear character definition.
  • No need for print heads or ribbons, reducing maintenance and operational costs.
  • Quiet operation and compact footprint suit space-limited lab environments.
  • Compatible with LIS/LIM systems for seamless integration.
Product Options:
  • Vega: Six cassette hoppers for multiple colors and styles.
  • Nova: Single hopper version for economical use.
Service & Support:
  • Includes 1-year base warranty + 2-year Total Care Plan (repairs, parts, and annual preventive maintenance).
  • Valid only with Epredia laser cassettes; third-party use reverts coverage to Premier Service.
Specifications:
  • Type: Laser Printer
  • Voltage: 100–240 V
  • Frequency: 50/60 Hz
  • Certifications: IEC 61010-2-101:2015, IEC 61010-1:2010, IEC 61326-1:2012, IEC 61326-2-6:2012
Specifications
Type Laser Printer
Voltage 100/240 V
Color Gray