Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vegetable Oil
SCI_ED

Catalog No. S802371 Shop All Fisherbrand Products
Change view
Click to view available options
Soybean Oil; 500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S802371 Soybean Oil; 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S802371

missing translation for 'validMainframeMsg'

Specifications

CAS 8001-22-7
Color Colorless
Packaging Soybean Oil
Quantity 500 mL
Synonym Soybean Oil
Chemical Name or Material Vegetable Oil
Safety handling Check Alert storage code Red
ShelfLife 36 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.